Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMF1
Entrez Gene ID
9491
Gene Name
proteasome inhibitor subunit 1
Gene Aliases
PI31
Chromosome Location
Chr.20: 1113229 - 1172246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323407.1 NP_001310336.1 2 - 3 330 78
NM_001323408.1 NP_001310337.1 2 - 3 459 78
NM_001323409.1 NP_001310338.1 2 - 3 459 78
NM_001323410.1 NP_001310339.1 2 - 3 459 78
NM_006814.4 NP_006805.2 2 - 3 459 78
NM_178578.3 NP_848693.2 3 - 4 480 78
GenBank mRNA AK294134.1 - 2 - 3 415 78
AK300667.1 - 2 - 3 204 78
AK302164.1 - 2 - 3 296 78
BC126462.1 - 3 - 4 336 78
BI461198.1 - 2 - 3 455 78
BI830805.1 - 3 - 4 413 78
D88378.1 - 2 - 3 404 78
DA949012.1 - 2 - 3 416 78
DB028624.1 - 3 - 4 446 78
DC398121.1 - 2 - 3 330 78
DC398668.1 - 2 - 3 459 78

View Full Product Details