Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ANAPC10
- Entrez Gene ID
-
10393
- Gene Name
- anaphase promoting complex subunit 10
- Gene Aliases
-
APC10, DOC1
- Chromosome Location
-
Chr.4: 144967117 - 145098541 on Build GRCh38
View Full Product Details