The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014934.4 | NP_055749.1 | 3 - 4 | 644 | 101 |
NM_198968.3 | NP_945319.1 | 3 - 4 | 644 | 101 | |
XM_005254032.1 | XP_005254089.1 | 3 - 4 | 644 | 101 | |
XM_006719916.1 | XP_006719979.1 | 3 - 4 | 644 | 101 | |
XM_006719917.2 | XP_006719980.1 | 3 - 4 | 644 | 101 | |
XM_006719918.1 | XP_006719981.1 | 3 - 4 | 644 | 101 | |
XM_006719920.1 | XP_006719983.1 | 3 - 4 | 644 | 101 | |
XM_017020444.1 | XP_016875933.1 | 3 - 4 | 644 | 101 | |
XM_017020445.1 | XP_016875934.1 | 3 - 4 | 644 | 101 | |
XM_017020446.1 | XP_016875935.1 | 3 - 4 | 644 | 101 | |
GenBank mRNA | AB023213.1 | - | 3 - 4 | 627 | 101 |
BC041804.1 | - | 3 - 4 | 644 | 101 | |
DR001764.1 | - | 3 - 4 | 564 | 101 |