The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282683.1 | NP_001269612.1 | 8 - 9 | 796 | 102 |
NM_024735.4 | NP_079011.3 | 7 - 8 | 1048 | 102 | |
GenBank mRNA | AF318348.1 | - | 6 - 7 | 1248 | 102 |
AF428140.1 | - | 7 - 8 | 1012 | 102 | |
AL117444.1 | - | 5 - 6 | 556 | 102 | |
AY736035.1 | - | 8 - 9 | 999 | 102 | |
BC012748.1 | - | 7 - 8 | 1022 | 102 |