The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002885.2 | NP_002876.2 | 1 - 2 | 111 | 78 |
XM_005245955.3 | XP_005246012.2 | 1 - 2 | 123 | 78 | |
XM_006710804.2 | XP_006710867.2 | 1 - 2 | 123 | 78 | |
XM_006710805.2 | XP_006710868.2 | 1 - 2 | 123 | 78 | |
XM_017001967.1 | XP_016857456.1 | 1 - 2 | 123 | 78 | |
XM_017001968.1 | XP_016857457.1 | 1 - 2 | 158 | 78 | |
XM_017001972.1 | XP_016857461.1 | 1 - 2 | 123 | 78 | |
XM_017001974.1 | XP_016857463.1 | 1 - 2 | 123 | 78 | |
XM_017001976.1 | XP_016857465.1 | 1 - 2 | 123 | 78 | |
XM_017001977.1 | XP_016857466.1 | 1 - 2 | 123 | 78 | |
GenBank mRNA | BP283134.1 | - | 1 - 2 | 111 | 78 |
M64788.1 | - | 1 - 2 | 54 | 78 |