Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAN2B2
Entrez Gene ID
23324
Gene Name
mannosidase alpha class 2B member 2
Gene Aliases
-
Chromosome Location
Chr.4: 6575174 - 6622403 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292038.1 NP_001278967.1 17 - 18 2698 144
NM_015274.2 NP_056089.1 17 - 18 2851 144
GenBank mRNA AK025803.1 - 9 - 10 1479 144
AK075246.1 - 5 - 6 789 144
AK301473.1 - 17 - 18 2683 144
AL832232.1 - 17 - 18 2821 144
AL833071.2 - 17 - 18 2824 144
BC033307.1 - 17 - 18 2820 144
BC036836.1 - 7 - 8 1033 144
BC094773.1 - 17 - 18 2851 144
BM730115.1 - 2 - 3 203 144

View Full Product Details