Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTPBP6
Entrez Gene ID
8225
Gene Name
GTP binding protein 6 (putative)
Gene Aliases
PGPL
Chromosome Location
Chr.X: 304750 - 318819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012227.3 NP_036359.3 7 - 8 1161 100
XM_006724868.3 XP_006724931.1 8 - 9 1061 100
XM_011545637.2 XP_011543939.1 6 - 7 766 100
GenBank mRNA AK024630.1 - 7 - 8 1100 100
AK026462.1 - 1 - 2 182 100
AK074528.1 - 1 - 2 1049 100
AK097872.1 - 2 - 3 1499 100
AK223412.1 - 7 - 8 1106 100
BC014636.1 - 7 - 8 834 100
BT007360.1 - 6 - 7 790 100
Y14391.2 - 7 - 8 1119 100

View Full Product Details