Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLK
Entrez Gene ID
51701
Gene Name
nemo like kinase
Gene Aliases
-
Chromosome Location
Chr.17: 28042647 - 28205140 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016231.4 NP_057315.3 10 - 11 1739 142
XM_005257988.2 XP_005258045.1 9 - 10 1667 142
GenBank mRNA AF180819.1 - 10 - 11 1737 142
AF197898.1 - 10 - 11 1739 142
AK024686.1 - 10 - 11 1371 142
AL161970.1 - 2 - 3 189 142
BC064663.1 - 10 - 11 2251 142
U69562.1 - 1 - 2 142

View Full Product Details