Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLK
Entrez Gene ID
51701
Gene Name
nemo like kinase
Gene Aliases
-
Chromosome Location
Chr.17: 28042647 - 28205140 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016231.4 NP_057315.3 4 - 5 961 81
XM_005257988.2 XP_005258045.1 4 - 5 976 81
GenBank mRNA AF180819.1 - 4 - 5 959 81
AF197898.1 - 4 - 5 961 81
AK024686.1 - 4 - 5 81
AK315315.1 - 4 - 5 976 81
BC064663.1 - 4 - 5 1473 81
U69562.1 - 7 - 8 81

View Full Product Details