The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016231.4 | NP_057315.3 | 6 - 7 | 1264 | 64 |
XM_005257988.2 | XP_005258045.1 | 6 - 7 | 1279 | 64 | |
GenBank mRNA | AF180819.1 | - | 6 - 7 | 1262 | 64 |
AF197898.1 | - | 6 - 7 | 1264 | 64 | |
AK024686.1 | - | 6 - 7 | 896 | 64 | |
AK315315.1 | - | 6 - 7 | 1279 | 64 | |
BC064663.1 | - | 6 - 7 | 1776 | 64 | |
U69562.1 | - | 5 - 6 | 64 |