The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199687.2 | NP_001186616.1 | 4 - 5 | 456 | 69 |
NM_001242310.1 | NP_001229239.1 | 3 - 4 | 358 | 69 | |
NM_001324206.1 | NP_001311135.1 | 4 - 5 | 458 | 69 | |
NM_021783.4 | NP_068555.1 | 4 - 5 | 458 | 69 | |
NR_136726.1 | 4 - 5 | 456 | 69 | ||
NR_136727.1 | 4 - 5 | 458 | 69 | ||
XM_011531000.1 | XP_011529302.1 | 4 - 5 | 408 | 69 | |
XM_011531001.1 | XP_011529303.1 | 4 - 5 | 456 | 69 | |
XM_011531002.1 | XP_011529304.1 | 4 - 5 | 408 | 69 | |
XM_011531004.1 | XP_011529306.1 | 4 - 5 | 408 | 69 | |
XM_011531006.2 | XP_011529308.1 | 4 - 5 | 448 | 69 | |
XM_011531007.2 | XP_011529309.1 | 4 - 5 | 448 | 69 | |
XM_017029704.1 | XP_016885193.1 | 4 - 5 | 408 | 69 | |
XM_017029706.1 | XP_016885195.1 | 4 - 5 | 408 | 69 | |
GenBank mRNA | AF298812.1 | - | 3 - 4 | 348 | 69 |
AK296264.1 | - | 4 - 5 | 408 | 69 | |
AK314881.1 | - | 4 - 5 | 424 | 69 | |
AY152724.1 | - | 3 - 4 | 348 | 69 | |
AY358735.1 | - | 3 - 4 | 351 | 69 | |
AY358736.1 | - | 3 - 4 | 351 | 69 | |
BC034919.1 | - | 4 - 5 | 405 | 69 | |
BP249106.1 | - | 4 - 5 | 424 | 69 | |
DA215848.1 | - | 4 - 5 | 457 | 69 | |
DA574050.1 | - | 4 - 5 | 408 | 69 |