Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZP2
Entrez Gene ID
7783
Gene Name
zona pellucida glycoprotein 2
Gene Aliases
ZPA, Zp-2
Chromosome Location
Chr.16: 21197452 - 21214510 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290104.1 NP_001277033.1 18 - 19 2470 73
NM_003460.2 NP_003451.1 18 - 19 2497 73
XM_011545942.2 XP_011544244.1 17 - 18 2059 73
GenBank mRNA AK310454.1 - 18 - 19 2497 73
AK313003.1 - 17 - 18 2039 73
BC096304.3 - 17 - 18 2014 73
BC096305.3 - 17 - 18 1987 73
BC096306.1 - 17 - 18 2014 73
BC096307.3 - 17 - 18 2014 73
M90366.1 - 17 - 18 2014 73

View Full Product Details