Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD1D
Entrez Gene ID
912
Gene Name
CD1d molecule
Gene Aliases
CD1A, R3, R3G1
Chromosome Location
Chr.1: 158178038 - 158186427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319145.1 NP_001306074.1 5 - 6 1207 83
NM_001766.3 NP_001757.1 6 - 7 1486 83
XM_005245583.1 XP_005245640.1 5 - 6 1165 83
XM_006711621.1 XP_006711684.1 5 - 6 1047 83
XM_011510125.1 XP_011508427.1 6 - 7 1301 83
XM_011510127.1 XP_011508429.1 5 - 6 1129 83
GenBank mRNA AK225996.1 - 4 - 5 921 83
BC027926.1 - 6 - 7 1486 83
J04142.1 - 5 - 6 1151 83

View Full Product Details