Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS3
Entrez Gene ID
10174
Gene Name
sorbin and SH3 domain containing 3
Gene Aliases
SCAM-1, SCAM1, SH3D4
Chromosome Location
Chr.8: 22544990 - 22575495 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005775.4 NP_005766.3 10 - 11 1157 122
XM_006716266.1 XP_006716329.1 11 - 12 1120 122
XM_006716268.2 XP_006716331.1 10 - 11 1521 122
GenBank mRNA AF037261.1 - 10 - 11 958 122
AK301196.1 - 9 - 10 876 122
AK303950.1 - 10 - 11 968 122
BC067260.1 - 10 - 11 943 122
BC091514.1 - 10 - 11 1157 122

View Full Product Details