Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACC1
Entrez Gene ID
6867
Gene Name
transforming acidic coiled-coil containing protein 1
Gene Aliases
Ga55
Chromosome Location
Chr.8: 38728186 - 38853028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005273625.4 XP_005273682.1 4 - 5 1887 71
XM_005273628.1 XP_005273685.1 3 - 4 1394 71
XM_005273629.1 XP_005273686.1 4 - 5 1407 71
XM_005273630.1 XP_005273687.1 3 - 4 1415 71
XM_011544631.1 XP_011542933.1 3 - 4 1773 71
XM_011544632.2 XP_011542934.1 5 - 6 1948 71
XM_011544633.1 XP_011542935.1 3 - 4 1773 71
XM_011544635.1 XP_011542937.1 3 - 4 1477 71
XM_017013770.1 XP_016869259.1 2 - 3 1307 71
XM_017013771.1 XP_016869260.1 2 - 3 1579 71
GenBank mRNA AB029026.1 - 1 - 2 307 71
AY039239.1 - 1 - 2 47 71

View Full Product Details