The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018704.2 | NP_061174.1 | 4 - 5 | 559 | 63 |
XM_011541781.2 | XP_011540083.1 | 4 - 5 | 630 | 63 | |
XM_017001806.1 | XP_016857295.1 | 4 - 5 | 997 | 63 | |
GenBank mRNA | AK022544.1 | - | 3 - 4 | 480 | 63 |
AL598321.1 | - | 4 - 5 | 515 | 63 | |
BC016029.1 | - | 4 - 5 | 499 | 63 |