The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290145.1 | NP_001277074.1 | 6 - 7 | 705 | 80 |
NM_015584.4 | NP_056399.1 | 6 - 7 | 759 | 80 | |
GenBank mRNA | AF077203.1 | - | 6 - 7 | 667 | 80 |
AF179891.1 | - | 6 - 7 | 618 | 80 | |
AK293707.1 | - | 6 - 7 | 579 | 80 | |
AK313232.1 | - | 6 - 7 | 651 | 80 | |
BC000655.2 | - | 6 - 7 | 651 | 80 | |
BC009265.2 | - | 6 - 7 | 645 | 80 | |
BC018864.2 | - | 6 - 7 | 684 | 80 |