Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAK1
Entrez Gene ID
578
Gene Name
BCL2 antagonist/killer 1
Gene Aliases
BAK, BAK-LIKE, BCL2L7, CDN1
Chromosome Location
Chr.6: 33572546 - 33580293 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC110337.1 - 4 - 5 559 64

View Full Product Details