Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZDHHC8
- Entrez Gene ID
-
29801
- Gene Name
- zinc finger DHHC-type containing 8
- Gene Aliases
-
DHHC8, ZDHHCL1, ZNF378
- Chromosome Location
-
Chr.22: 20131841 - 20148007 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB033118.1 |
- |
1 - 2 |
153 |
75 |
View Full Product Details