The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000185.3 | NP_000176.2 | 1 - 2 | 50 | 117 |
GenBank mRNA | BP237845.1 | - | 1 - 2 | 54 | 117 |