Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPIND1
Entrez Gene ID
3053
Gene Name
serpin family D member 1
Gene Aliases
D22S673, HC2, HCF2, HCII, HLS2, LS2, THPH10
Chromosome Location
Chr.22: 20774095 - 20787720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000185.3 NP_000176.2 3 - 4 1224 66
GenBank mRNA AK225292.1 - 3 - 4 1207 66
AK314200.1 - 3 - 4 1209 66
BC035028.2 - 3 - 4 1206 66
CR456573.1 - 2 - 3 1289 66
M12849.1 - 2 - 3 1185 66
X03498.1 - 2 - 3 1102 66

View Full Product Details