The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193489.1 | NP_001180418.1 | 14 - 15 | 2280 | 128 |
NM_014701.3 | NP_055516.2 | 13 - 14 | 2145 | 128 | |
GenBank mRNA | AK307480.1 | - | 14 - 15 | 1963 | 128 |
BC033001.2 | - | 14 - 15 | 2215 | 128 | |
D87445.3 | - | 13 - 14 | 2015 | 128 |