The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018090.6 | NP_001018100.1 | 7 - 8 | 937 | 110 |
NM_001018091.6 | NP_001018101.1 | 7 - 8 | 937 | 110 | |
NM_001285900.3 | NP_001272829.1 | 7 - 8 | 937 | 110 | |
NR_104367.2 | 7 - 8 | 937 | 110 | ||
NR_104368.2 | 7 - 8 | 937 | 110 | ||
NR_104369.2 | 7 - 8 | 937 | 110 | ||
NR_104370.2 | 7 - 8 | 937 | 110 | ||
NR_104371.3 | 7 - 8 | 937 | 110 | ||
GenBank mRNA | AY207007.1 | - | 7 - 8 | 808 | 110 |
AY208913.1 | - | 7 - 8 | 808 | 110 | |
AY331564.1 | - | 7 - 8 | 808 | 110 | |
AY333779.1 | - | 7 - 8 | 808 | 110 | |
AY334560.1 | - | 7 - 8 | 808 | 110 | |
AY334561.1 | - | 7 - 8 | 808 | 110 | |
AY334562.1 | - | 6 - 7 | 715 | 110 | |
AY341345.1 | - | 5 - 6 | 601 | 110 | |
AY353057.1 | - | 7 - 8 | 808 | 110 | |
AY353058.1 | - | 7 - 8 | 808 | 110 | |
AY353060.1 | - | 6 - 7 | 715 | 110 | |
JF419331.1 | - | 7 - 8 | 827 | 110 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018100.4 | NP_001018110.1 | 7 - 8 | 937 | 110 |
NM_152451.7 | NP_689664.3 | 7 - 8 | 937 | 110 | |
GenBank mRNA | AK055535.1 | - | 7 - 8 | 900 | 110 |
AL833028.1 | - | 2 - 3 | 180 | 110 | |
AL833142.1 | - | 6 - 7 | 718 | 110 | |
AY207459.1 | - | 7 - 8 | 808 | 110 | |
AY237639.1 | - | 7 - 8 | 808 | 110 | |
BC101645.1 | - | 7 - 8 | 937 | 110 | |
BC112148.1 | - | 7 - 8 | 935 | 110 | |
FJ970029.1 | - | 7 - 8 | 813 | 110 |