Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP3K1
Entrez Gene ID
4214
Gene Name
mitogen-activated protein kinase kinase kinase 1
Gene Aliases
MAPKKK1, MEKK, MEKK 1, MEKK1, SRXY6
Chromosome Location
Chr.5: 56815073 - 56896152 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005921.1 NP_005912.1 1 - 2 981 168
GenBank mRNA AF042838.1 - 1 - 2 423 168

View Full Product Details