The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080435.2 | NP_001073904.1 | 5 - 6 | 1370 | 89 |
XM_005272421.4 | XP_005272478.1 | 5 - 6 | 986 | 89 | |
XM_005272422.3 | XP_005272479.1 | 5 - 6 | 733 | 89 | |
XM_005272423.4 | XP_005272480.1 | 5 - 6 | 972 | 89 | |
XM_011521232.2 | XP_011519534.1 | 3 - 4 | 1182 | 89 | |
XM_017021923.1 | XP_016877412.1 | 5 - 6 | 2819 | 89 | |
GenBank mRNA | AB075851.1 | - | 5 - 6 | 1356 | 89 |
AK126887.1 | - | 5 - 6 | 1371 | 89 |