The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000964.3 | NP_000955.1 | 6 - 7 | 1285 | 68 |
NM_001024809.3 | NP_001019980.1 | 5 - 6 | 1478 | 68 | |
NM_001145301.2 | NP_001138773.1 | 6 - 7 | 1397 | 68 | |
NM_001145302.2 | NP_001138774.1 | 4 - 5 | 1106 | 68 | |
XM_005257552.4 | XP_005257609.1 | 5 - 6 | 855 | 68 | |
XM_005257553.1 | XP_005257610.1 | 6 - 7 | 1189 | 68 | |
XM_005257554.1 | XP_005257611.1 | 6 - 7 | 1438 | 68 | |
XM_011525095.1 | XP_011523397.1 | 7 - 8 | 1335 | 68 | |
XM_011525096.1 | XP_011523398.1 | 5 - 6 | 648 | 68 | |
XM_017024920.1 | XP_016880409.1 | 5 - 6 | 757 | 68 | |
GenBank mRNA | AK098172.1 | - | 5 - 6 | 857 | 68 |
AK130192.1 | - | 6 - 7 | 1321 | 68 | |
AK292205.1 | - | 6 - 7 | 1337 | 68 | |
AK312564.1 | - | 6 - 7 | 1261 | 68 | |
BC008727.2 | - | 6 - 7 | 1262 | 68 | |
BC071733.1 | - | 5 - 6 | 1081 | 68 | |
FJ487576.1 | - | 4 - 5 | 1042 | 68 | |
HQ692826.1 | - | 5 - 6 | 807 | 68 | |
HQ692827.1 | - | 5 - 6 | 792 | 68 | |
S50916.1 | - | - | 669 | 68 | |
X06538.1 | - | 5 - 6 | 1033 | 68 | |
X06614.1 | - | 5 - 6 | 909 | 68 |