Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RARA
Entrez Gene ID
5914
Gene Name
retinoic acid receptor alpha
Gene Aliases
NR1B1, RAR
Chromosome Location
Chr.17: 40309171 - 40357643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011525095.1 XP_011523397.1 2 - 3 167 77

View Full Product Details