The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321079.1 | NP_001308008.1 | 11 - 12 | 1497 | 88 |
NM_001321081.1 | NP_001308010.1 | 12 - 13 | 1561 | 88 | |
NM_001321082.1 | NP_001308011.1 | 11 - 12 | 1468 | 88 | |
NM_001991.4 | NP_001982.2 | 12 - 13 | 1588 | 88 | |
XM_005257145.2 | XP_005257202.1 | 10 - 11 | 1343 | 88 | |
XM_011524517.2 | XP_011522819.1 | 11 - 12 | 1436 | 88 | |
XM_017024350.1 | XP_016879839.1 | 11 - 12 | 1415 | 88 | |
XM_017024351.1 | XP_016879840.1 | 10 - 11 | 1322 | 88 | |
XM_017024352.1 | XP_016879841.1 | 9 - 10 | 1173 | 88 | |
GenBank mRNA | AB002386.1 | - | 11 - 12 | 1500 | 88 |
AB004818.1 | - | 11 - 12 | 1506 | 88 | |
AK295626.1 | - | 11 - 12 | 1450 | 88 | |
AK295853.1 | - | 10 - 11 | 1288 | 88 | |
AK299887.1 | - | 11 - 12 | 1403 | 88 | |
AK304835.1 | - | 11 - 12 | 1401 | 88 | |
AK309121.1 | - | 11 - 12 | 1408 | 88 | |
AK310399.1 | - | 10 - 11 | 1280 | 88 | |
AK310556.1 | - | 4 - 5 | 632 | 88 | |
AL833159.1 | - | 12 - 13 | 1508 | 88 | |
BC015882.1 | - | 12 - 13 | 1522 | 88 | |
BT009782.1 | - | 10 - 11 | 1400 | 88 | |
L38934.1 | - | 5 - 6 | 604 | 88 | |
U50315.1 | - | 11 - 12 | 1506 | 88 |