Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EZH1
Entrez Gene ID
2145
Gene Name
enhancer of zeste 1 polycomb repressive complex 2 subunit
Gene Aliases
KMT6B
Chromosome Location
Chr.17: 42700275 - 42745096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321079.1 NP_001308008.1 15 - 16 1937 89
NM_001321081.1 NP_001308010.1 16 - 17 2001 89
NM_001321082.1 NP_001308011.1 15 - 16 1908 89
NM_001991.4 NP_001982.2 16 - 17 2028 89
XM_005257145.2 XP_005257202.1 14 - 15 1783 89
XM_011524517.2 XP_011522819.1 15 - 16 1876 89
XM_017024350.1 XP_016879839.1 15 - 16 1855 89
XM_017024351.1 XP_016879840.1 14 - 15 1762 89
XM_017024352.1 XP_016879841.1 13 - 14 1613 89
GenBank mRNA AB002386.1 - 15 - 16 1940 89
AB004818.1 - 15 - 16 1943 90
AK295626.1 - 15 - 16 1890 89
AK295853.1 - 14 - 15 1728 89
AK299887.1 - 15 - 16 1843 89
AK304835.1 - 15 - 16 1841 89
AK309121.1 - 15 - 16 1848 89
AK310399.1 - 14 - 15 1720 89
AK310556.1 - 8 - 9 1072 89
AL833159.1 - 16 - 17 1948 89
BC015882.1 - 16 - 17 1962 89
BT009782.1 - 14 - 15 1840 89
U50315.1 - 15 - 16 1946 89

View Full Product Details