Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPA13
Entrez Gene ID
6782
Gene Name
heat shock protein family A (Hsp70) member 13
Gene Aliases
STCH
Chromosome Location
Chr.21: 14371115 - 14383188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006948.4 NP_008879.3 1 - 2 96 60
GenBank mRNA AK128540.1 - 1 - 2 54 60
AK223282.1 - 1 - 2 54 60
AK299004.1 - 1 - 2 45 60
AK312396.1 - 1 - 2 51 60
BC036370.2 - 1 - 2 58 60
U04735.1 - 1 - 2 63 60

View Full Product Details