The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176820.3 | NP_789790.2 | 1 - 2 | 310 | 94 |
GenBank mRNA | AY154464.1 | - | 1 - 2 | 282 | 94 |
BC136583.1 | - | 1 - 2 | 302 | 94 |