The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270622.1 | NP_001257551.1 | 1 - 2 | 192 | 122 |
NM_001270623.1 | NP_001257552.1 | 1 - 2 | 192 | 122 | |
XM_011538989.2 | XP_011537291.1 | 1 - 2 | 298 | 122 | |
GenBank mRNA | DR000391.1 | - | 1 - 2 | 201 | 122 |