The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270622.1 | NP_001257551.1 | 4 - 5 | 664 | 79 |
NM_001270623.1 | NP_001257552.1 | 4 - 5 | 684 | 79 | |
NM_004731.4 | NP_004722.2 | 3 - 4 | 522 | 79 | |
NR_073055.1 | 5 - 6 | 798 | 79 | ||
NR_073056.1 | 5 - 6 | 659 | 79 | ||
XM_005269231.4 | XP_005269288.1 | 3 - 4 | 458 | 79 | |
XM_011538989.2 | XP_011537291.1 | 5 - 6 | 868 | 79 | |
XM_011538990.2 | XP_011537292.1 | 4 - 5 | 760 | 79 | |
XM_011538991.2 | XP_011537293.1 | 4 - 5 | 558 | 79 | |
XM_011538992.2 | XP_011537294.1 | 4 - 5 | 1493 | 79 | |
XM_011538993.2 | XP_011537295.1 | 6 - 7 | 1258 | 79 | |
XM_011538995.2 | XP_011537297.1 | 6 - 7 | 1253 | 79 | |
XM_017020224.1 | XP_016875713.1 | 7 - 8 | 1179 | 79 | |
XM_017020225.1 | XP_016875714.1 | 7 - 8 | 1172 | 79 | |
XM_017020226.1 | XP_016875715.1 | 7 - 8 | 1307 | 79 | |
XM_017020227.1 | XP_016875716.1 | 6 - 7 | 853 | 79 | |
GenBank mRNA | AF049608.1 | - | 4 - 5 | 519 | 79 |
AF058056.1 | - | 3 - 4 | 485 | 79 | |
AK301725.1 | - | 5 - 6 | 786 | 79 | |
AK310281.1 | - | 5 - 6 | 626 | 79 | |
AK313345.1 | - | 3 - 4 | 492 | 79 | |
BC030693.2 | - | 3 - 4 | 514 | 79 |