The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175053.3 | NP_778223.2 | 2 - 3 | 721 | 106 |
GenBank mRNA | AJ508777.1 | - | 2 - 3 | 721 | 106 |