Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM172A
Entrez Gene ID
83989
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
Chromosome Location
Chr.5: 93615130 - 94111699 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163417.1 NP_001156889.1 2 - 3 295 125
NM_001163418.1 NP_001156890.1 2 - 3 295 125
NM_032042.5 NP_114431.2 3 - 4 419 125
XM_005272103.3 XP_005272160.1 3 - 4 385 125
XM_005272105.3 XP_005272162.1 3 - 4 384 125
XM_005272106.3 XP_005272163.1 3 - 4 385 125
XM_005272109.4 XP_005272166.1 3 - 4 385 125
XM_005272110.3 XP_005272167.1 2 - 3 239 125
XM_005272111.4 XP_005272168.1 3 - 4 385 125
XM_006714717.3 XP_006714780.1 3 - 4 411 125
XM_011543668.2 XP_011541970.1 3 - 4 385 125
XM_011543670.2 XP_011541972.1 3 - 4 385 125
XM_011543671.2 XP_011541973.1 3 - 4 385 125
XM_011543672.2 XP_011541974.1 3 - 4 315 125
XM_011543674.2 XP_011541976.1 3 - 4 385 125
XM_017009952.1 XP_016865441.1 3 - 4 707 125
XM_017009953.1 XP_016865442.1 3 - 4 705 125
XM_017009954.1 XP_016865443.1 3 - 4 385 125
XM_017009955.1 XP_016865444.1 2 - 3 1123 125
XM_017009956.1 XP_016865445.1 2 - 3 437 125
XM_017009959.1 XP_016865448.1 2 - 3 464 125
XM_017009961.1 XP_016865450.1 3 - 4 329 125
XM_017009962.1 XP_016865451.1 2 - 3 251 125
GenBank mRNA AK295879.1 - 2 - 3 244 125
AK296986.1 - 2 - 3 272 125
AK312930.1 - 3 - 4 324 125
AL136630.1 - 3 - 4 258 125
BC020584.2 - 2 - 3 221 125

View Full Product Details