Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FAM172A
- Entrez Gene ID
-
83989
- Gene Name
- family with sequence similarity 172 member A
- Gene Aliases
-
C5orf21
- Chromosome Location
-
Chr.5: 93615130 - 94111699 on Build GRCh38
View Full Product Details