Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM172A
Entrez Gene ID
83989
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
Chromosome Location
Chr.5: 93615130 - 94111699 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163417.1 NP_001156889.1 8 - 9 1098 67
NM_001163418.1 NP_001156890.1 7 - 8 906 67
NM_032042.5 NP_114431.2 9 - 10 1222 67
NR_028080.1 7 - 8 929 67
XM_005272103.3 XP_005272160.1 9 - 10 1188 67
XM_005272105.3 XP_005272162.1 8 - 9 995 67
XM_005272106.3 XP_005272163.1 8 - 9 969 67
XM_005272108.3 XP_005272165.1 8 - 9 1116 67
XM_005272110.3 XP_005272167.1 7 - 8 823 67
XM_006714717.3 XP_006714780.1 9 - 10 1214 67
XM_006714718.3 XP_006714781.1 7 - 8 1828 67
XM_006714719.3 XP_006714782.1 7 - 8 977 67
XM_011543668.2 XP_011541970.1 9 - 10 1188 67
XM_011543670.2 XP_011541972.1 9 - 10 1188 67
XM_011543671.2 XP_011541973.1 9 - 10 1188 67
XM_011543672.2 XP_011541974.1 9 - 10 1118 67
XM_011543673.2 XP_011541975.1 8 - 9 1017 67
XM_017009952.1 XP_016865441.1 9 - 10 1510 67
XM_017009953.1 XP_016865442.1 9 - 10 1508 67
XM_017009954.1 XP_016865443.1 9 - 10 1188 67
XM_017009955.1 XP_016865444.1 8 - 9 1926 67
XM_017009956.1 XP_016865445.1 8 - 9 1240 67
XM_017009957.1 XP_016865446.1 7 - 8 1194 67
XM_017009958.1 XP_016865447.1 7 - 8 989 67
XM_017009960.1 XP_016865449.1 6 - 7 771 67
GenBank mRNA AK295127.1 - 8 - 9 1121 67
AK295879.1 - 7 - 8 855 67
AK296986.1 - 8 - 9 1075 67
AK297471.1 - 6 - 7 691 67
AK312930.1 - 9 - 10 1127 67
BC020584.2 - 8 - 9 1024 67

View Full Product Details