The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014902.5 | NP_055717.2 | 7 - 8 | 2143 | 110 |
XM_011528688.2 | XP_011526990.1 | 6 - 7 | 1909 | 110 | |
XM_011528689.2 | XP_011526991.1 | 6 - 7 | 1916 | 110 | |
GenBank mRNA | AB023181.1 | - | 7 - 8 | 2085 | 110 |