The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242692.1 | NP_001229621.1 | 15 - 16 | 2247 | 69 |
NM_007163.3 | NP_009094.3 | 14 - 15 | 2729 | 69 | |
XM_017026015.1 | XP_016881504.1 | 16 - 17 | 2295 | 69 | |
XM_017026016.1 | XP_016881505.1 | 14 - 15 | 2034 | 69 | |
GenBank mRNA | AF349446.1 | - | - | 2729 | 69 |
AK292422.1 | - | 15 - 16 | 2234 | 69 | |
BC110445.1 | - | 13 - 14 | 1937 | 69 | |
BC110446.1 | - | 13 - 14 | 1937 | 69 |