The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014321.3 | NP_055136.1 | 6 - 7 | 690 | 73 |
NR_037620.1 | 6 - 7 | 749 | 73 | ||
GenBank mRNA | AF139658.1 | - | 6 - 7 | 678 | 73 |
AK021946.1 | - | 6 - 7 | 651 | 73 | |
AK024019.1 | - | 6 - 7 | 684 | 73 | |
BC039032.1 | - | 6 - 7 | 670 | 73 | |
BC063565.1 | - | 6 - 7 | 659 | 73 |