The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007014.4 | NP_008945.2 | 2 - 3 | 156 | 78 |
XM_017022880.1 | XP_016878369.1 | 3 - 4 | 192 | 78 | |
GenBank mRNA | AJ001982.1 | - | 1 - 2 | 84 | 78 |
U96114.1 | - | 2 - 3 | 88 | 78 |