The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020958.2 | NP_066009.2 | 1 - 2 | 273 | 104 |
NM_058237.1 | NP_478144.1 | 1 - 2 | 273 | 104 | |
XM_017021529.1 | XP_016877018.1 | 1 - 2 | 246 | 104 | |
XM_017021530.1 | XP_016877019.1 | 1 - 2 | 231 | 104 | |
GenBank mRNA | AB046842.1 | - | 1 - 2 | 192 | 104 |
BC002650.2 | - | 1 - 2 | 247 | 104 | |
BC068491.1 | - | 1 - 2 | 232 | 104 | |
BI546744.1 | - | 1 - 2 | 277 | 105 |