The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_058237.1 | NP_478144.1 | 20 - 21 | 2347 | 71 |
XM_005267930.2 | XP_005267987.1 | 20 - 21 | 2180 | 71 | |
XM_011537038.2 | XP_011535340.1 | 20 - 21 | 2343 | 71 | |
XM_011537039.2 | XP_011535341.1 | 19 - 20 | 2088 | 71 | |
XM_011537040.2 | XP_011535342.1 | 18 - 19 | 1995 | 71 | |
XM_017021528.1 | XP_016877017.1 | 18 - 19 | 2032 | 71 | |
XM_017021529.1 | XP_016877018.1 | 22 - 23 | 2702 | 71 | |
XM_017021530.1 | XP_016877019.1 | 22 - 23 | 2676 | 71 | |
GenBank mRNA | AB046842.1 | - | 20 - 21 | 2266 | 71 |
AK303722.1 | - | 8 - 9 | 829 | 71 | |
BC068491.1 | - | 20 - 21 | 2306 | 71 |