Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SERPINA5
Entrez Gene ID
5104
Gene Name
serpin family A member 5
Gene Aliases
PAI-3, PAI3, PCI, PCI-B, PLANH3, PROCI
Chromosome Location
Chr.14: 94581369 - 94593120 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000624.5 NP_000615.3 1 - 2 120 59

View Full Product Details