The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000624.5 | NP_000615.3 | 3 - 4 | 858 | 94 |
GenBank mRNA | AK096131.1 | - | 3 - 4 | 563 | 94 |
AK293192.1 | - | 4 - 5 | 553 | 94 | |
AK298279.1 | - | 3 - 4 | 464 | 94 | |
AK310869.1 | - | 2 - 3 | 733 | 94 | |
AL080185.1 | - | 1 - 2 | 542 | 94 | |
BC008915.2 | - | 3 - 4 | 753 | 94 | |
J02639.1 | - | 2 - 3 | 669 | 94 | |
U35464.1 | - | 1 - 2 | 647 | 94 |