The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005268098.4 | XP_005268155.1 | 1 - 2 | 107 | 104 |
XM_011537177.2 | XP_011535479.1 | 1 - 2 | 107 | 104 | |
XM_011537179.2 | XP_011535481.1 | 1 - 2 | 107 | 104 | |
XM_017021660.1 | XP_016877149.1 | 1 - 2 | 119 | 104 | |
XM_017021661.1 | XP_016877150.1 | 1 - 2 | 171 | 104 | |
XM_017021662.1 | XP_016877151.1 | 1 - 2 | 99 | 104 | |
GenBank mRNA | DB484577.1 | - | 1 - 2 | 171 | 104 |