The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322405.1 | NP_001309334.1 | 6 - 7 | 910 | 74 |
NM_001322406.1 | NP_001309335.1 | 7 - 8 | 1026 | 74 | |
NM_001322408.1 | NP_001309337.1 | 5 - 6 | 830 | 74 | |
NM_001322410.1 | NP_001309339.1 | 6 - 7 | 914 | 74 | |
NM_001322412.1 | NP_001309341.1 | 6 - 7 | 910 | 74 | |
NM_001322413.1 | NP_001309342.1 | 5 - 6 | 834 | 74 | |
NM_001322414.1 | NP_001309343.1 | 6 - 7 | 914 | 74 | |
NM_001322415.1 | NP_001309344.1 | 5 - 6 | 834 | 74 | |
NM_001322416.1 | NP_001309345.1 | 6 - 7 | 914 | 74 | |
NM_015491.2 | NP_056306.1 | 5 - 6 | 834 | 74 | |
NM_032870.3 | NP_116259.2 | 6 - 7 | 914 | 74 | |
NR_136326.1 | 5 - 6 | 834 | 74 | ||
XM_005266912.3 | XP_005266969.2 | 5 - 6 | 793 | 74 | |
XM_017010710.1 | XP_016866199.1 | 6 - 7 | 911 | 74 | |
XM_017010711.1 | XP_016866200.1 | 5 - 6 | 797 | 74 | |
XM_017010712.1 | XP_016866201.1 | 6 - 7 | 2265 | 74 | |
XM_017010713.1 | XP_016866202.1 | 6 - 7 | 2261 | 74 | |
GenBank mRNA | AF314184.1 | - | 4 - 5 | 704 | 74 |
AF314186.1 | - | 6 - 7 | 852 | 74 | |
AK027658.1 | - | 6 - 7 | 877 | 74 | |
AK027759.1 | - | 7 - 8 | 853 | 74 | |
AK027898.1 | - | 6 - 7 | 870 | 74 | |
AK074628.1 | - | 5 - 6 | 797 | 74 | |
AK291155.1 | - | 4 - 5 | 683 | 74 | |
BC007791.2 | - | 6 - 7 | 847 | 74 | |
BC012780.2 | - | 6 - 7 | 856 | 74 | |
BC019074.2 | - | 6 - 7 | 823 | 74 | |
BC024655.1 | - | 1 - 2 | 278 | 74 | |
BC042850.1 | - | 7 - 8 | 1023 | 74 | |
BC052638.1 | - | 6 - 7 | 852 | 74 | |
BC064413.1 | - | 6 - 7 | 848 | 74 | |
BC064640.1 | - | 6 - 7 | 827 | 74 |