Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP2R5C
Entrez Gene ID
5527
Gene Name
protein phosphatase 2 regulatory subunit B'gamma
Gene Aliases
B56G, PR61G
Chromosome Location
Chr.14: 101760573 - 101927992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161725.1 NP_001155197.1 4 - 5 475 83
NM_001161726.1 NP_001155198.1 4 - 5 547 83
NM_002719.3 NP_002710.2 2 - 3 426 83
NM_178586.2 NP_848701.1 2 - 3 426 83
NM_178587.2 NP_848702.1 2 - 3 426 83
XM_005267819.1 XP_005267876.1 4 - 5 536 83
XM_005267820.1 XP_005267877.1 4 - 5 476 83
XM_005267822.3 XP_005267879.1 4 - 5 536 83
XM_005267823.1 XP_005267880.1 4 - 5 536 83
XM_005267824.1 XP_005267881.1 4 - 5 536 83
XM_005267826.1 XP_005267883.1 2 - 3 249 83
XM_005267827.1 XP_005267884.1 4 - 5 471 83
XM_011536914.2 XP_011535216.1 3 - 4 340 83
XM_011536915.2 XP_011535217.1 3 - 4 423 83
XM_011536916.2 XP_011535218.1 3 - 4 1422 83
XM_011536918.1 XP_011535220.1 3 - 4 577 83
XM_011536919.2 XP_011535221.1 3 - 4 559 83
XM_011536920.1 XP_011535222.1 3 - 4 453 83
XM_011536921.2 XP_011535223.1 2 - 3 342 83
XM_017021422.1 XP_016876911.1 3 - 4 492 83
XM_017021423.1 XP_016876912.1 2 - 3 426 83
XM_017021424.1 XP_016876913.1 3 - 4 453 83
GenBank mRNA AB451341.1 - 2 - 3 286 83
AI692258.1 - 4 - 5 548 83
AK131391.1 - 4 - 5 460 83
AK302470.1 - 4 - 5 375 83
AK315072.1 - 2 - 3 362 83
BC016183.1 - 2 - 3 324 83
BI560029.1 - 2 - 3 400 83
D26445.1 - 2 - 3 283 83
DA853338.1 - 2 - 3 426 83
L42375.1 - 2 - 3 256 83
U37352.1 - 2 - 3 344 83
Z69030.1 - 2 - 3 364 83

View Full Product Details