Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADGRL2
Entrez Gene ID
23266
Gene Name
adhesion G protein-coupled receptor L2
Gene Aliases
CIRL2, CL2, LEC1, LPHH1, LPHN2
Chromosome Location
Chr.1: 81306155 - 81992738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005270666.4 XP_005270723.1 8 - 9 2714 81
XM_006710485.3 XP_006710548.1 9 - 10 2730 81
XM_006710488.2 XP_006710551.1 9 - 10 2024 81
XM_006710489.3 XP_006710552.1 9 - 10 1915 81
XM_017000782.1 XP_016856271.1 13 - 14 2598 81
XM_017000783.1 XP_016856272.1 10 - 11 2009 81
XM_017000784.1 XP_016856273.1 9 - 10 1914 81
XM_017000785.1 XP_016856274.1 9 - 10 2021 81
XM_017000786.1 XP_016856275.1 9 - 10 2025 81
XM_017000787.1 XP_016856276.1 9 - 10 1936 81
XM_017000790.1 XP_016856279.1 10 - 11 1950 81
XM_017000791.1 XP_016856280.1 8 - 9 2014 81
XM_017000793.1 XP_016856282.1 10 - 11 1952 81
XM_017000794.1 XP_016856283.1 10 - 11 1951 81
XM_017000795.1 XP_016856284.1 10 - 11 1950 81
XM_017000796.1 XP_016856285.1 8 - 9 1974 81
GenBank mRNA AB018329.1 - 3 - 4 465 81

View Full Product Details