The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017922.1 | NP_001017922.1 | 9 - 10 | 922 | 107 |
NM_018538.3 | NP_061008.2 | 8 - 9 | 818 | 107 | |
XM_006710313.3 | XP_006710376.1 | 10 - 11 | 1187 | 107 | |
XM_011540570.2 | XP_011538872.1 | 9 - 10 | 878 | 107 | |
GenBank mRNA | AF311285.1 | - | 8 - 9 | 818 | 107 |
AK056138.1 | - | 9 - 10 | 896 | 107 | |
AK314436.1 | - | 9 - 10 | 906 | 107 | |
AY049028.1 | - | 8 - 9 | 836 | 107 | |
BC099703.1 | - | 8 - 9 | 751 | 107 | |
BC099707.1 | - | 8 - 9 | 756 | 107 | |
BC099712.1 | - | 8 - 9 | 751 | 107 | |
BC099713.1 | - | 8 - 9 | 751 | 107 | |
BX537371.1 | - | 6 - 7 | 1420 | 107 |